Alkaline detergent no foam, RBS 50
Leverandør: CHEMICAL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RBS50/E123 RBS50/E148
CHEPRBS50/E123EA 1290 DKK
CHEPRBS50/E123 CHEPRBS50/E148
Alkaline detergent no foam, RBS 50
Renseopløsninger Detergenter og Overflade Rengøringsmidler
Efficient cleaning of glassware and laboratory equipment in automatic washing machines - labwashers.
  • High detergency power
  • Contains active chlorine
  • Low use concentration
  • Easy rinse
RBS 50 is recommended for cleaning glassware and laboratory equipment in automatic washing machine - labwashers. It efficiently removes organic and biological residues, distilation residues etc.
Bestillingsoplysninger: Available in box of 4 × 5 litres; 20 kg drum. Please contact Avantor for details.
Advarsel: Not recommended for use on white metals.