Alkaline detergent no foam, RBS 48 S
Leverandør: CHEMICAL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
48S/E123 RBS48SE030
CHEP48S/E123EA 821 DKK
CHEP48S/E123 CHEPRBS48SE030
Alkaline detergent no foam, RBS 48 S
Renseopløsninger Detergenter og Overflade Rengøringsmidler
Efficient cleaning of glassware and laboratory equipment in automatic washing machines - labwashers.
  • High detergency power
  • Low use concentration
  • Easy rinse
RBS 48 S allows cleaning and degreasing of glassware and laboratory equipment. It removes organic and biological residues. Compatible with use in hard water. Can also be used in ultrasonic cleaner.
Bestillingsoplysninger: Available in box of 4 × 5 litres; 220 kg drum.
Advarsel: Not recommended for use on white metals.