Dehydrated culture media, YPD (YEPD) media, MP Biomedicals
Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
114001232 114001222 114001032 114001022 114001036 114001042
ICNA114001232EA 1740 DKK
ICNA114001232 ICNA114001222 ICNA114001032 ICNA114001022 ICNA114001036 ICNA114001042
Dehydrated culture media, YPD (YEPD) media, MP Biomedicals
Mikrobiologiske medier Dehydreret Mikrobiologisk medie
YPD Broth is used for routine growth of Saccharomyces cerevisiae for molecular biology studies.
  • Available in different packaging format - pre-measured capsules and bulk powder packing
  • YPD agar media contains agar in addition to the other ingredients
It is a nutritionally rich medium containing dextrose, peptone and yeast extract. Dextrose is the energy source. Yeast extract and peptone provide carbon, nitrogen, minerals, vitamins, trace ingredients and other essential growth nutrients which stimulate the growth of yeasts.