Bottles, extra wide neck, round with screw cap
Leverandør: MURRAY S
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CO60UC20 CO250UC10 CO120UC20
215-2191EA 437 DKK
215-2191 215-2189 215-1919
Bottles, extra wide neck, round with screw cap
Flasker Glasflasker
Glass, clear, with black urea screw cap.
  • Good chemical and physical properties
  • For use in applications that are not associated with high thermal loads
  • Stable and robust
These bottles are specially suited for dry chemicals and specimens of all descriptions, but can also be used for liquids. The wide opening enables bottles to be filled conveniently and safely with powders, pastes, granulates and liquids.