Cleanroom face masks with visors, BioClean Clearview™, PE/PP
Leverandør: Nitritex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VFM210-L VFM210-T-SLOT
113-2266EA 5020 DKK
113-2266 NITRVFM210-T-SLOT
Cleanroom face masks with visors, BioClean Clearview™, PE/PP
Masker Renrumsmasker
Face masks made from 3 ply, PP/PE, ultrasonically bonded fabric with integrated anti-fog visors. The outer facing consists of hydrophobic non woven PE, the filter layer of meltblown PE and the inner facing of hydrophilic PP/PE (Non woven).
  • No latex proteins
  • Particle filtration efficiency (PFE): >97%, bacterial filtration efficiency (BFE): >96,5%
  • Barcodes appear on the outer carton to allow automatic data entry into stock control systems
  • With ties or ear loops
Suitable for use in class 10, ISO 4, M2.5 environments.
Thickness (visor): 0,18 mm