Cleanroom lab coats, BioClean-D™, CleanTough™, disposable
Leverandør: Nitritex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BDLC-L BDLC-M BDLC-S BDLC-XL BDLC-XXL S-BDLC-M S-BDLC-S S-BDLC-XL S-BDLC-XXL S-BDLC-L
113-0809EA 2920 DKK
113-0809 113-0810 113-0811 113-0812 113-0813 113-0825 113-0826 113-0827 113-0828 113-0824
Cleanroom lab coats, BioClean-D™, CleanTough™, disposable
Engangsbeklædning
These disposable lab coats are made from CleanTough™ fabric which is spunbonded non-woven PP laminated with a film of PE. The fabric provides protection against fine sprays and particles.
  • Lightweight, low linting, and hard-wearing material
  • Three pockets, press stud fastening
  • Open cuffs and rear vent
  • Available as sterile or non sterile version
  • A unique barcode on the outer carton (ITF-14) allows automatic data entry into Stock Control Systems
Suitable for use in Class 10, ISO 4, M2.5 environments.
Colour: White
Type PB [6]; Cat. III
Certificeringer: EN ISO 13034 (Type PB[6]), EN 14325, EN 1149-5, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2; Cat. III
Manufactured in a facility holding ISO 9001:2008 and 89/686/EEC Article 11b certifications.
Sterile models: Sterilised in accordance with ISO 11137-1:2006.
Emballering: One lab coat per sealed double plastic bag; 30 plastic bags per sealed carton liner; one carton liner per carton (30 lab coats).