Tychem® 2000 C og Tychem® 6000 F kemisk resistente forklæder
Leverandør: DuPont
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TYCPA30SYL00 TYFPA30SGY00
113-0875EA 1740 DKK
113-0875 113-0880
Tychem® 2000 C og Tychem® 6000 F kemisk resistente forklæder
Arbejdstøj
Specially designed for use with Tychem® apparel, these aprons offer enhanced protection for body parts that are more exposed to hazardous substances.
  • Shin-length apron with neck and waist ties
  • Anti-static treatment (EN 1149-5) on inside
  • Conform to EN 14126 (barrier to infective agents)
Tychem® 2000 C aprons use the strength of Tyvek®, and a polymeric barrier coating, to offer good permeation protection against a wide range of inorganic chemicals and biological hazards (even under pressure). They are used for splash or pressurised splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing.
Tychem® 6000 F aprons are made from a lightweight, durable fabric consisting of a proprietary barrier film laminated to a heavy duty Tyvek® substrate. They offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards, and certain chemical warfare agents. These aprons are used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.
Colour: Yellow (Tychem® 2000 C) or grey (Tychem® 6000 F)
Type PB [3], EN 14126, Cat. III
Certificeringer: Type PB [3], EN 14126, kat. III