Cut resistant gloves, AlphaTec® 16-650
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
16-650/10 16-650/7 16-650/8 16-650/9
112-3777EA 105 DKK
112-3777 112-3774 112-3775 112-3776
Cut resistant gloves, AlphaTec® 16-650
Handsker
Raised crinkle cut finish made from natural rubber on interlock knitted liner with protective cuff.
  • Increased protection against sharp edges
  • Soft knitted cotton lining provides good padding and cut resistance with a high level of comfort
  • Raised crinkle finish delivers an excellent grip on slippery surfaces in both wet and dry conditions
  • Good anatomical design shape gives improved fit and increased safety
Length: 310 mm
Colour: Green
EN 388 (X241), EN 407 (X2XXXX), EN 374, Cat. III
Certificeringer: EN 388:2016 (X141B), EN 407 (X2XXXX), EN ISO 374-1:2016 Type B (KLT), EN ISO 374-5:2016, Cat. III