Chemical resistant gowns, Tychem® 2000 C and Tychem® 6000 F, model PL50
Leverandør: DuPont
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TYCPL50SYLLXXL TF0290TGY00L2X TF0290TGY00SM TF0290TGY0034X
113-0876EA 0 DKK
113-0876 DUPOTF0290TGY00L2X DUPOTF0290TGY00SM DUPOTF0290TGY0034X
Chemical resistant gowns, Tychem® 2000 C and Tychem® 6000 F, model PL50
Engangsbeklædning
Shin-length gown with wrap-over rear closure, hook and loop neck closure, and waist ties.
  • Elasticated wrists
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Anti-static treatment (EN 1149-5) on inside
Tychem® 2000 C garments utilise the strength of Tyvek® and a polymeric barrier coating to offer good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure). They are used for splash or pressurised splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing.
Tychem® 6000 F garments are made from a lightweight and durable fabric consisting of a proprietary barrier film laminated to a heavy duty Tyvek® substrate. They offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents. Tychem® F is used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.
Colour: Yellow (Tychem® 2000 C) or grey (Tychem® 6000 F)
Weight: 83 g/m² (Tychem® 2000 C); 120 g/m² (Tychem® 6000 F)
Certificeringer: Type PB[3], EN 14126; Cat. III