Dehydrated culture media, superbroth, Molecular Biology Certified™, MP Biomedicals
Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113010031 113010032 113010036
ICNA113010031EA 1930 DKK
ICNA113010031 ICNA113010032 ICNA113010036
Dehydrated culture media, superbroth, Molecular Biology Certified™, MP Biomedicals
Mikrobiologiske medier Dehydreret Mikrobiologisk medie
Superbroth is a rich medium with high levels of tryptone and yeast extract used for obtaining high yields of E. coli cells and lambda bacteriophages.
  • Available as capsules or powder
  • Molecular biology certified
Due to the high levels of peptides, amino acids, vitamins and cofactors high yields of lambda phage and phagemids in E. coli are obtained.