Wipes and wipe rolls for general applications, WYPALL* L20 EXTRA / L20 EXTRA+
Leverandør: KIMBERLY CLARK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7300 7301 7303 7302 7308 7248 7317 7356
115-2206EA 425 DKK
115-2206 115-0024 115-2208 115-2207 115-2013 115-1936 115-1543 KIMB7356
Wipes and wipe rolls for general applications, WYPALL* L20 EXTRA / L20 EXTRA+
Papirservietter Engangsservietter og - håndklæder Papirservietter til alment brug
These single-use wipes, made from two ply AIRFLEX* material, are ideal for multipurpose cleaning and medium duty wiping tasks such as polishing glass, smooth surfaces and cleaning tools.
  • Stronger than conventional two ply wipes
  • Strong and absorbent
  • Clean up fast without disintegrating
  • Can be used with a dispensing system to help control use and reduce wastage
Small rolls: For use with dispenser 7041.
Large rolls: For use with dispensers 6146, 6154 and 6155.
Centrefeed rolls: For use with dispenser 7017.