VWR® Durac™ Plus, Hydrometers
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-2388EA 1640 DKK
620-2388 620-2389
VWR® Durac™ Plus, Hydrometers
Flydevægte
These plain form hydrometers are designed for applications where rough handling or contamination could be an issue.
  • Durable and shatterproof, lead free ballast
  • Easy to read black scale on yellow back to avoid eye fatigue
  • Individually serialised - Temperature standardisation of 15 °C
Suitable for applications in the food or beverage industries.
Certificeringer: NIST.
Bestillingsoplysninger: Includes a multi-language statement of accuracy.
Advarsel: Not recommended for use with petroleum-based liquids or other corrosives, these should be storedMore Product Information