Cryoboxes, PP
Leverandør: RATIOLAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5320410 5320413 5320313 5320314 5320315 5320316
RATI5320410EA 316 DKK
RATI5320410 RATI5320413 RATI5320313 RATI5320314 RATI5320315 RATI5320316
Cryoboxes, PP
Bokse Æsker til Kryorør
Boxes made of high quality PP.
  • Autoclavable
  • Temperature-resistant from –90 to +121 °C
  • Numerically coded
  • Very good chemical resistance
With an numerical coding and corresponding vents for draining condensed water. The boxes are adjustable in height.