VWR®, Calibrated Electronic Verification Thermometers
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-0904EA 1050 DKK
620-0904 620-2148 620-2344
VWR®, Calibrated Electronic Verification Thermometers
Termometre Digital termometre
Digital thermometers verify sample temperatures in freezers, refrigerators, incubators and ovens. Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample.
  • Individually serialised
Plastic bottles cannot be used in ovens.
Certificeringer: ISO 17025, ISO/IEC 17025:2005, traceable to NIST.
Leveringsinformation: Includes multi-language instructions (English, French, German, Italian, Portuguese and Spanish), AAA or LR44 (620-2148) batteries.
Advarsel: * Internal
** External