VWR®, Calibrated Electronic Thermometers with Stainless Steel Probe
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-0916EA 517 DKK
620-0916 620-2343 620-2709 620-2710
VWR®, Calibrated Electronic Thermometers with Stainless Steel Probe
Termometre Digital termometre
These high accuracy digital thermometers, with chemical resistant probes, monitor the temperature of pharmaceuticals, vaccines, chemicals, blood and blood products, food products and more.
  • Features include: 10-event recording, max./min. temperature readings, timer and alarm
  • Calibration points: 0; 100 °C
Certificeringer: Traceable to NIST, individually serialised, CE marked, ISO/IEC 17025:2005.
Leveringsinformation: Includes accredited calibration documentation and multi-language instructions (English, French, German, Italian, Portuguese and Spanish) and AAA battery.