Earmuffs, Peltor™ Optime™ II
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H520B H520F H520A H520P3E1 H520P3E H520P3EV H520BGQ
111-0455EA 436 DKK
111-0455 111-0456 111-0454 111-1079 111-1078 111-1082 MMMAH520BGQ
Earmuffs, Peltor™ Optime™ II
Høreværn Støjblokerende høreværn
Optime™ II earmuffs are available in various designs for different requirements: With foldable headband, neckband or as a helmet attachment.
  • Generous inside depth for added comfort
  • Soft, wide ear cushions filled with a unique fluid/foam combination ensure very low contact pressure
  • Ear cushions feature ventilation channels and are covered with soft, hygienic foil
  • Two-point fasteners made of rust-proof stainless steel ensure low contact pressure even during longer periods of use
Ideal for use in noisy industrial environments, e.g. for construction machine operators or airport personnel.
SNR: 31 dB (headband and neckband); 30 dB (helmet-mounted)
Advarsel: *Suitable for all Peltor™ helmets, except G2000