Industrial grade cleanroom tapes, EGV™
Leverandør: MICRONOVA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EGV-1WH_U EGV-2OR EGV-2RE EGV-34RE
819-0027EA 0 DKK
819-0027 MICREGV-2OR MICREGV-2RE MICREGV-34RE
Industrial grade cleanroom tapes, EGV™
Tape
These industrial grade cleanroom tapes are made of vinyl with rubber adhesive.
  • Light industrial use
  • Clean construction
  • Forms strong bond without leaving adhesive residue
  • Can be written on with cleanroom pen
A tape for cleanroom support applications. Processed in a controlled environment. Wound on a plastic core and sleeve bagged.
Length: 32,9 m
Different colours available.
Certificeringer: Complies with Federal Specification PPP-T-66E and CID A-A 1689A.