Extreme temperature cleanroom tapes
Leverandør: MICRONOVA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
825-14AM 825-12AM 825-1AM 825-3AM_U 825-34AM 825-38AM_U 825-15AM_U
817-0055EA 269 DKK
817-0055 817-0054 817-0057 819-0072 819-0070 819-0071 819-0067
Extreme temperature cleanroom tapes
Tape
These extreme temperature cleanroom tapes are made of polyamide film with silicone adhesive.
  • Flame resistant electrical insulation
  • Solder mask for circuit boards
  • Heat sealing and lamination
  • Cold storage
  • Lyophilising and freeze drying
Polyamide film with pressure-sensitive silicone adhesive. The extreme heat resistant properties (−73 to +260 °C) of this tape make it suitable for high heat and sub zero applications. The silicone adhesive ensures a clean release from most substrates even after being exposed to extreme temperatures.
Length: 32,9 mm
Colour: Amber transparent