Conductive grid tapes, CGT™
Leverandør: MICRONOVA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CGT-12 CGT-34_U CGT-2 CGT-6 CGT-1 CGT-15 CGT-38
819-0074EA 167 DKK
819-0074 819-0077 819-0076 819-0079 819-0018 819-0075 819-0078
Conductive grid tapes, CGT™
Tape
Conductive, anti-static polypropylene tape, specially designed with conductive grids embedded between two anti-static layers, for use on devices in ESD sensitive environments.
  • Heavy-duty, anti-static copolymer tape with acrylic adhesive
  • Manufactured for use in ESD sensitive environments
  • Features a conductive grid embedded between two antistatic layers
  • Conducts charge away from substrate
  • Protects sensitive devices that would be otherwise damaged by normal insulating tape
  • Provides an effective static shield and prevents static buildup
  • Prevents shedding, flaking, cracking, or chipping of the grids
  • 36 yards wound on a 3" plastic core
The Micronova CGT™ Conductive Grid Tape features conductive grids that are embedded between two anti-static layers to provide an effective static shield. This design prevents static build-up, as well as preventing any shedding, flaking, cracking, or chipping of the grids.
Bestillingsoplysninger: For additional widths, contact your Avantor sales representative.