Zinkoxid Ph. Eur., Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Zinci oxidum, Zinc (II) oxide

P106-33EA 343 DKK
P106-33 P106-42
Zinkoxid Ph. Eur., Macron Fine Chemicals™
Zinkoxid

Certificeringer: Meets Ph. Eur. Chemical Specifications.

Formel: ZnO
Molekylvægt: 81.39 g/mol
MDL Number: MFCD00011300
CAS-nummer: 1314-13-2
UN nummer: 3077
ADR: 9 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR