Drug test kit
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
321481PEA 8470 DKK
321481P 321602G 321612Y 321622K 321632M 321642X 321652Q
Drug test kit
Narkotikatest
This kit provides a convenient means of screening suspect material for restricted drugs. The scheme and reagents were developed at the laboratory of the government chemist. Each kit provides sufficient reagents for at least 200 tests.
  • Kittet består af følgende:
  • 20 (4 boxes of 5, 0.2 ml) ampoules of marquis reagent
  • 1×10 ml kokain reagens
  • 1×10 ml barbiturat reagens
  • 1×10 ml LSD reagens
  • 1×20 ml cannabis reagens 1
  • 1×10 ml cannabis reagens 2
  • 1×5 g cannabis special solid reagens
  • 1x20 g neutralisator
  • 4 boxes of Whatman filterpapir
  • 1 mikro-spatel
  • 20 ampul breakers