Capillary columns, BP624
Leverandør: Trajan Scientific and Medical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
054827 054832 054835 054836 054842 054841 054840 054838
568-0268EA 6210 DKK
568-0268 558-0337 568-0272 558-0338 568-0274 568-0273 558-0339 558-0340
Capillary columns, BP624
Kromatografikolonner
A polar phase used for EPA volatile organics analysis (EPA 624, SW-846 8240/8260), alcohol analysis and for USP G43.
Cyanopropylphenyl polysiloxane.
Maximum temperatures: 230/240 °C
Suitable replacement for: DB-624, Rtx Volatiles, VOCOL, OV-624, AT-624, HP-624, 007-624, VB-624, ZB-624, HP-VOC, CP-Select624CB.