Anti-NR3C2 Rabbit Polyclonal Antibody (Cy5.5®)

Leverandør: Bioss
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BS-1850R-CY5.5
BOSSBS-1850R-CY5.5EA 6790 DKK
BOSSBS-1850R-CY5.5
Anti-NR3C2 Rabbit Polyclonal Antibody (Cy5.5®)
Antistoffer
Receptor for both mineralocorticoids (MC) such as aldosterone and glucocorticoids (GC) such as corticosterone or cortisol. Binds to mineralocorticoid response elements (MRE) and transactivates target genes. The effect of MC is to increase ion and water transport and thus raise extracellular fluid volume and blood pressure and lower potassium levels.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NR3C2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR