Anti-TNNT2 Rabbit Polyclonal Antibody (Cy7®)

Leverandør: Bioss
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BS-10648R-CY7
BOSSBS-10648R-CY7EA 6590 DKK
BOSSBS-10648R-CY7
Anti-TNNT2 Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
Troponin T is the tropomyosin-binding subunit of troponin, the thin filament regulatory complex which confers calcium-sensitivity to striated muscle actomyosin ATPase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Troponin T
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR