Eyepiece cameras, Dino-Eye
Leverandør: Dino-Lite / IDCP B.V.
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AM4023CT AM4023U AM7025X
630-1473EA 2270 DKK
630-1473 630-1474 630-2922
Eyepiece cameras, Dino-Eye
Kameraer
With these cameras users can turn their conventional microscopes into digital microscopes.The ocular (eyepiece) of the non-digital microscope is simply replaced by an Dino-Eye eyepiece and connected to a computer via USB.
  • Different models for different eyepiece sizes
  • High speed USB 2.0 connection
  • Software included for image or video processing, incl. calibrated measurement features
  • MicroTouch feature (model AM4023CT only): Touch sensitive area that users can use to capture images instead of using a keyboard and mouse
  • Model AM7025X: Edge series with