Sealing rings for dessicators
Leverandør: KLEINFELD
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1213642
467-2240EA 1800 DKK
467-2240
Sealing rings for dessicators
Forsegling Tætningsringe
Soft rubber sealing rings, makes grease unnecessary. Suitable for desiccators and Witt jars.