VWR®, Measuring Beakers with Handle, PMP TPX
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
213-0504EA 22.4 DKK
213-0504 213-0505 213-0506 213-0507 213-0508 213-0509
VWR®, Measuring Beakers with Handle, PMP TPX
Bægre
Made of PMP, crystal-clear.
  • With printed graduations
  • Excellent chemical resistance
  • With handle for better usage
  • Withstand temperatures of 0 to 121 °C when used continuously (up to 170 °C for short periods)
  • Resin meets US FDA 21 CFR requirements