VWR®, Measuring Cylinders, Tall Form, Class B, PMP TPX
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
612-5599EA 140 DKK
612-5599 612-5589 612-5587 612-5573 612-5596 612-5572 612-5601 612-5579
VWR®, Measuring Cylinders, Tall Form, Class B, PMP TPX
Cylindere Målecylindre
PMP (TPX), crystal-clear, tall form, class B.
  • Stable hexagonal base
  • Graduations are calibrated in mould for accuracy and consistency
  • Excellent chemical resistance
  • Withstand temperatures of 0 to 121 °C when used continuously (up to 170 °C short period)
  • Resin meets US FDA 21 CFR requirements
Certificeringer: DIN 12681/ISO 6706