Instrument trays
Leverandør: Sampling Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A674-215 A674-295 A674-395 A674-490
SAMPA674-215EA 239 DKK
SAMPA674-215 SAMPA674-295 SAMPA674-395 SAMPA674-490
Instrument trays
Bakker/Tallerkener/Potter Instrumentbakker/-skåle
Made from 304 stainless steel.
  • Good chemical resistance, very good temperature resistance
  • Crevice free design
  • High quality polished finish
Ideal for general laboratory use as well as for carrying surgical instruments, sampling tools etc.