Funnels, with short stem, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6120P-50 6120P-40 6120P-30 6120P-150 6120P-120 6120P-75 6120P-100
CORN6120P-50EA 258 DKK
CORN6120P-50 CORN6120P-40 CORN6120P-30 CORN6120P-150 CORN6120P-120 CORN6120P-75 CORN6120P-100
Funnels, with short stem, Corning®
Tragte Filtreringstragte Filtertragte
Funnels, made of durable PP, translucent.
  • Very good chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper
Allow safe and easy transfers of solids or liquids from one vessel to another.