Jars, wide mouth, PFA
Leverandør: SAVILLEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
100-0060-01 100-0360-01 100-2000-01
SAVI100-0060-01EA 335 DKK
SAVI100-0060-01 SAVI100-0360-01 SAVI100-2000-01
Jars, wide mouth, PFA
Dåser Dåser, allround
Jars, made of molded translucent PFA.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Screw caps available seperately
For sample collection, storage and transport. With radiused inside design for easy sample removal.