Busulphan ≥98%, crystalline solid

Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1,4-Butanediol dimethanesulphonate, Butane-1,4-diyl dimethanesulphonate

14843-25 14843-50 14843-100 14843-250
CAYM14843-25EA 1030 DKK
CAYM14843-25 CAYM14843-50 CAYM14843-100 CAYM14843-250
Busulphan ≥98%, crystalline solid
Busulphan

1,4-Butanediol Dimethanesulfonate (Busulfan) is an alkyl sulfonate that acts as an alkylating antineoplastic agent. It forms both intra- and interstrand crosslinks on DNA. In mammals, busulfan causes profound and prolonged reduction in the generation of hematopoietic progenitors without significantly affecting lymphocyte levels and humoral antibody responses.


  • Soluble in DMSO and dimethylformamide
  • Stable for atleast two years


Synonyms: Busulphan, Mielosan, Milecitan, Myeloleukon, Mylecytan, Myleran, NCI C01592, NSC 750

Formel: C₆H₁₄O₆S₂
Molekylvægt: 246.29 g/mol
Smeltepunkt: 114…118°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007562
CAS-nummer: 55-98-1
UN nummer: 2811
ADR: 6.1 I

Bestil nu

Specifikationer Testresultater

Purity ≥98%
Stability ≥2 years at −20 °C
Form Crystalline solid
Solubility Sparingly soluble in aqueous buffers
Storage temperature −20 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR