VWR®, Reagent Bottles, Narrow Neck, with Standard Ground Stoppers
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
215-3859EA 174 DKK
215-3859 215-3860 215-3861 215-3862 215-3863 215-3864 215-3865 215-3866
VWR®, Reagent Bottles, Narrow Neck, with Standard Ground Stoppers
Flasker Glasflasker
Borosilicate 3.3 glass according to ISO 4796.
  • High resistance to chemicals and thermal shock
  • Available in capacities from 10 to 2000 ml
  • Hollow glass stopper and ground joint
These reagent bottles have a precise ground neck to ensure a water- and air-tight connection between the bottle and the glass stopper. All components are made from borosilicate 3.3 glass and are, therefore, ideal for the storage of aggressive and hot media.