Nicotinamid-adenin dinucleotid (oxideret form) 93
Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Nadide, NAD+, Nicotinamide adenine dinucleotide, oxidized
0210031905 0210031901 0210031925
ICNA0210031905EA 1590 DKK
ICNA0210031905 ICNA0210031901 ICNA0210031925
Nicotinamid-adenin dinucleotid (oxideret form) 93
Nicotinamid-adenin dinucleotid (oxideret form)
This grade of NAD is intended for applications where maximum reagent purity is not critical. Well suited for use on most automatic analysers.
  • White powder, soluble in water (200 mg/ml)
  • pH: 2 - 4 (1% aqueous solution)
Solutions should be aliquoted and stored at –20 °C for up to one week.
Formel: C₂₁H₂₇N₇O₁₄P₂
Molekylvægt: 663.43 g/mol
MDL Number: MFCD00036253
CAS-nummer: 53-84-9
Merck Index: 13,06370