DeepWell™ plates, 96 square well, Nunc™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
95040452 95040462
NUNC95040452EA 2910 DKK
NUNC95040452 735-0398
DeepWell™ plates, 96 square well, Nunc™
Mikroplader Deepwell Mikroplader
Ideal for sample collection, storage and combinatorial chemistry library applications.
  • Square well shape reduces liquid retention
  • Resistant to most chemicals, solvents and alcohols used in combinatorial chemistry
  • Sterile format certified free from RNase