Tray, Rotilabo®
Leverandør: Roth Carl
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8103.1 8104.1 8105.1 8106.1
ROTH8103.1EA 0 DKK
ROTH8103.1 ROTH8104.1 ROTH8105.1 ROTH8106.1
Tray, Rotilabo®
Bakker/Tallerkener/Potter Laboratoriebakker/-skåle
Dishes, made of stainless steel, chrome-nickel plated.
  • With folding handles on two sides
  • Very good temperature resistance
  • Easy to clean
  • Autoclavable
Suitable for food applications.