Cut resistant gloves, HyFlex® 11-724
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11-724/9 11-724/10 11-724/8 11-724/11 11-724/7 11-724/6
ANSE11-724/9EA 1230 DKK
ANSE11-724/9 ANSE11-724/10 ANSE11-724/8 ANSE11-724/11 ANSE11-724/7 ANSE11-724/6
Cut resistant gloves, HyFlex® 11-724
Handsker
These knitted seamless liners with INTERCEPT™ technology yarn and PU coated palm combine excellent flexibility and fit with optimal cut protection.
  • Best-in-class cut protection and increased comfort and dexterity
  • Palm coating design improves worker grip and extends glove life in abrasive conditions
  • Uncoated back allows the hand to breath
  • Grey coating hides dirt in environments with dirt exposure
INTERCEPT™ is a specialised knitting technology which blends engineered, synthetic and natural fibres into high performance yarns. This provides high-cut protection with exceptional comfort and dexterity.
Ideal for assembling metal parts and components, light metal presses, maintenance, plastics injection and moulding, and primary assembly.
Certificeringer: EN 388:2016 (4342B); Cat. II