Flame retardant and chemical resistant overalls, MICROCHEM® CFR, model 111
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RD96T-00111-03 RD96T-00111-04 RD96T-00111-05 RD96T-00111-06 RD96T-00111-07
ANSERD96T-00111-03EA 11080 DKK
ANSERD96T-00111-03 ANSERD96T-00111-04 ANSERD96T-00111-05 ANSERD96T-00111-06 ANSERD96T-00111-07
Flame retardant and chemical resistant overalls, MICROCHEM® CFR, model 111
Engangsbeklædning
These overalls are made from a flame retardant material designed to be worn over woven thermal protective garments such as NOMEX® or PYROVATEX®, offering protection from particulates and pressurised liquid spray without compromising worker protection in the event of a flash fire.
  • Two-piece hood
  • Elasticated hood, wrists and ankles
  • Double zip closure
  • Stitched and taped seams increased strength and provide an effective liquid and particle barrier
  • Flame retardant treated fabric with PVC barrier film offering protection from liquid chemicals to EN Type 3 & 4
  • Highly visible bright red colour to improve worker safety
MICROCHEM® CFR offers wearers protection from liquid chemicals to EN Types 3 & 4 and peace of mind to workers in potentially explosive/flammable environments, by decreasing the risk of burn injury when worn over thermal protective workwear.
Typical applications include: Oil and petrochemicals, petroleum distribution and processing, utilities.
Certificeringer: Type 3/4/5, EN ISO 14116 (index 1/0/0), EN 1073-2, EN 1149-5; Cat. III
Advarsel: MICROCHEM® CFR should never be worn in isolation for flame retardant protection. Always wear over the top of garments which achieve EN ISO 14116 Index 2 or above.