Chemical resistant overalls, with integrated socks, Microchem® 4000, model 122
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
GR40T-00122-02 GR40T-00122-03 GR40T-00122-04 GR40T-00122-05 GR40T-00122-06 GR40T-00122-07
ANSEGR40T-00122-02EA 5760 DKK
ANSEGR40T-00122-02 ANSEGR40T-00122-03 ANSEGR40T-00122-04 ANSEGR40T-00122-05 ANSEGR40T-00122-06 ANSEGR40T-00122-07
Chemical resistant overalls, with integrated socks, Microchem® 4000, model 122
Engangsbeklædning
These overalls are made of multi-layer barrier fabric (external film barrier, non woven inner layer, core high barrier film). They are designed to provide an exceptional barrier against organic and inorganic chemicals and biological agents.
  • Double zip system helps ensure a liquid tight seal without the need for additional taping
  • Double cuffs with knitted inner cuff
  • Elasticated 2-piece hood, outer cuffs, waist and boot overflaps
  • Integrated socks with boot overflap
  • Ultrasonically welded and taped seams
  • Textile-like inner improves wearer acceptance
  • Permeation tested against over 180 chemicals, including chemical warfare agents
Typical applications include: Chemical handling, transportation, oil-based mud protection, HAZMAT emergency response, pharmaceutical, mining and agriculture.
Certificeringer: Type 3/4/5, EN 14126, EN 1073-2, EN 1149-5; Cat. III
Suitable for Ex-Zones.