Chemical resistant overalls, with attached gloves, Microchem® 4000, model 121
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
GR40T-00121-03 GR40T-00121-04 GR40T-00121-05 GR40T-00121-06 GR40T-00121-07
ANSEGR40T-00121-03EA 9110 DKK
ANSEGR40T-00121-03 ANSEGR40T-00121-04 ANSEGR40T-00121-05 ANSEGR40T-00121-06 ANSEGR40T-00121-07
Chemical resistant overalls, with attached gloves, Microchem® 4000, model 121
Engangsbeklædning
These overalls are made of multi-layer barrier fabric (external film barrier, non woven inner layer, core high barrier film). They are designed to provide an exceptional barrier against organic and inorganic chemicals and biological agents.
  • Double zip system helps ensure a liquid tight seal without the need for additional taping
  • Elasticated 2-piece hood, waist and ankles
  • Attached Ansell Barrier® gloves
  • Ultrasonically welded and taped seams
  • Textile-like inner improves wearer acceptance
  • Permeation tested against over 180 chemicals, including chemical warfare agents
Typical applications include: Chemical handling, transportation, oil-based mud protection, HAZMAT emergency response, pharmaceutical, mining and agriculture. Also suitable for use in EX zones.
Certificeringer: Type 3/4/5, EN 14126, EN 1073-2, EN 1149-5; Cat. III
Suitable for Ex-Zones