Chemical resistant overalls, Microchem® 4000
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
GR40T-00111-02 GR40T-00111-03 GR40T-00111-04 GR40T-00111-05 GR40T-00111-06 GR40T-00111-07 GR40T-00103-04 GR40T-00103-05 GR40T-00103-06 GR40T-00103-07 GR40T-00103-03
ANSEGR40T-00111-02EA 4720 DKK
ANSEGR40T-00111-02 ANSEGR40T-00111-03 ANSEGR40T-00111-04 ANSEGR40T-00111-05 ANSEGR40T-00111-06 ANSEGR40T-00111-07 ANSEGR40T-00103-04 ANSEGR40T-00103-05 ANSEGR40T-00103-06 ANSEGR40T-00103-07 ANSEGR40T-00103-03
Chemical resistant overalls, Microchem® 4000
Engangsbeklædning
These overalls are made of multi-layer barrier fabric (external film barrier, non woven inner layer, core high barrier film). They are designed to provide an exceptional barrier against organic and inorganic chemicals and biological agents.
  • With collar or elasticated 2-piece hood
  • Double zip system helps ensure a liquid tight seal without the need for additional taping
  • Double cuffs with knitted inner cuff
  • Elasticated outer cuffs, waist and ankles
  • Ultrasonically welded and taped seams
  • Textile-like inner improves wearer acceptance
  • Permeation tested against over 180 chemicals, including chemical warfare agents
Typical applications include: Chemical handling, transportation, oil-based mud protection, HAZMAT emergency response, pharmaceutical, mining and agriculture. Also suitable for use in EX zones.
Certificeringer: Type 3/4/5, EN 14126, EN 1073-2, EN 1149-5; Cat. III
Suitable for Ex-Zones.