HPLC maintenance kits
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
25097 25322 25325 26548
REST25097EA 6000 DKK
REST25097 REST25322 REST25325 REST26548
HPLC maintenance kits
Kromatografi forsyninger Kromatografi reservedele
These kits are used for start-up and maintenance in all HPLC systems.
PEEK kits contain tubing, fittings, and tools essential for setting up and maintaining your HPLC system: PEEK tubing, connectors, and elbows, PTFE tubing, a tubing cutter and extra blades, a ValvTool wrench, open-end wrenches and more.
Stainless steel kits contain a wide range of tubing, fittings, and tools necessary to set up and maintain your HPLC system: a selection of lengths and IDs of ¹/₁₆" tubing, plus nuts, ferrules, a ValvTool wrench, and a zero-dead-volume union.