Nuts and ferrules, Rheodyne®
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
25096 25095
REST25096EA 0 DKK
REST25096 REST25095
Nuts and ferrules, Rheodyne®
Fittings og stik
Replacement long nut connects stainless steel tubing to a Rheodyne® 6-port valve or other Rheodyne® part.