Column end-fittings
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
25077 25078
REST25077EA 326 DKK
REST25077 REST25078
Column end-fittings
Fittings og stik
These ¹/₄" compression end-fittings are compatible with ¹/₁₆" connecting tubing using Valco and Parker fittings.
  • Fittings with distribution cone are intended for use on 3,0 to 4,6 mm ID HPLC columns
  • Fittings with flat bottom are intended for use on 1,0 to 3,0 mm ID HPLC columns