Industrial overalls, AlphaTec® 1500 PLUS STITCHED, model 111
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BL15S-00111-05 BL15S-00111-02 BL15S-00111-03 BL15S-00111-06 BL15S-00111-07 BL15S-00111-04 WH15S-00111-02 WH15S-00111-03 WH15S-00111-04 WH15S-00111-05 WH15S-00111-06 WH15S-00111-07 BL15S-00111-08 BL15S-00111-09
ANSEBL15S-00111-05EA 1700 DKK
ANSEBL15S-00111-05 113-9971 113-9972 ANSEBL15S-00111-06 113-9973 ANSEBL15S-00111-04 113-9974 113-9975 113-9976 113-9977 113-9978 113-9979 ANSEBL15S-00111-08 ANSEBL15S-00111-09
Industrial overalls, AlphaTec® 1500 PLUS STITCHED, model 111
Engangsbeklædning
These overalls are made from a highly breathable anti-static SMS fabric which utilises the latest developments in micro-fibre technology to ensure good filtration efficiency.
  • Proven to filter 99,9% of particles >3 µm (JSTIIF particle penetration test)
  • Air and water vapour permeable ('breathable') to help reduce the risk of heat stress
  • Silicone-free: Critical in spray painting applications
  • Optimised body fit improves wearer comfort and safety
  • Elasticated 3-piece hood, wrists, waist and ankles
  • Two-way front zipper with resealable storm flap
  • Stitched seams: Combining strength with particle barrier
  • Finger loops
SMS fabrics are a particularly good barrier against particulates such as asbestos, brick dust and cement dust, and will provide protection from light aerosol mists; as found in some paint spray environments.
Typical applications include: Asbestos related work, handling powders, general maintenance, construction, pharmaceutical industries, wood and metal processing, touch-up paint spraying and fibreglass, resin applications and ceramic fibres.
Weight: 48 to 60 g/m²
Certificeringer: Type 5/6, EN 1073-2, EN 1149-5; Cat. III.