Industrial overalls, AlphaTec® 2000 COMFORT, model 129
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
WH20B-00129-02 WH20B-00129-03 WH20B-00129-04 WH20B-00129-05 WH20B-00129-06 WH20B-00129-07
ANSEWH20B-00129-02EA 2520 DKK
ANSEWH20B-00129-02 ANSEWH20B-00129-03 ANSEWH20B-00129-04 ANSEWH20B-00129-05 ANSEWH20B-00129-06 ANSEWH20B-00129-07
Industrial overalls, AlphaTec® 2000 COMFORT, model 129
Engangsbeklædning
The critical areas to the front of the garment (including the hood, arms and legs) are made from the MICROGARD® 2000 STANDARD fabric (PE laminate with non woven inner layer), offering a high level of liquid and particle protection with a low level of water vapour resistance. The back panel is made from breathable MICROGARD® 1500 PLUS fabric (SMS) which is air and water vapour permeable.
  • Elasticated 3-piece hood, wrists, waist and ankles
  • Two-way front zipper with resealable storm flap; finger loops
  • Breathable SMS back panel allows airflow around the suit, increasing wearer comfort
  • Bound seams for superior strength, liquid and particle barrier
  • Air and water vapour permeable ('breathable') to help reduce the risk of heat stress
  • Silicone-free: Critical in spray painting applications
MICROGARD® 2000 COMFORT has been specifically designed for those working in warmer climates or warm working environments to help reduce the risk of heat stress.
Typical application includes: Pharmaceutical industry, cleanrooms, paint spraying, veterinary services and pest control.
Air permeability (back panel) result according to EN ISO 9237 of 160 L/m².s
Water vapour resistance according to EN 31092 of Ret <15*
Weight: 53 g/m²
Certificeringer: Type 5/6, EN 1073-2, EN 1149-5; Cat. III