Raw polyurethane foam (PUF) plugs and glass holder
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
22957 22965
REST22957EA 411 DKK
REST22957 REST22965
Raw polyurethane foam (PUF) plugs and glass holder
Prøvetagere Luftprøvetager
PUF plugs fits to glass holders as an alternative to pre-cleaned packed cartridges.
  • Small glass holder with a stem
  • Tubing connection to sampling pump
  • Durable and reusable PUF glass holders
Designed for air sampling applications.