Zwitterionic columns, CHIRALPAK® ZWIX
Leverandør: CHIRAL TECHNOLOGIES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
51345 51335 52335 52345
554-4594EA 134730 DKK
554-4594 554-4593 554-4595 554-4596
Zwitterionic columns, CHIRALPAK® ZWIX
Kromatografikolonner
The CHIRALPAK® ZWIX columns were developed for separation of a great variety of zwitterionic molecules, specifically, for separation of underivatised amino acids.
ZWIX selectors are zwitterionic molecules that incorporate both anion- and cation-exchange functional groups. These ZWIX selectors exhibit exceptional enantioseparation capabilities toward zwitterionic molecules such as amino acids and small peptides.