In-line filters, UHPLC, UltraLine
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
24993 24994
REST24993EA 1990 DKK
REST24993 REST24994
In-line filters, UHPLC, UltraLine
Filtre Indsatsfiltre
Stainless steel filter (0,5 µm) in a stainless steel housing. Its design easily installs with standard fittings.
  • Economical protection for UHPLC systems
  • Reliable way to extend column lifetime
  • Pressure rated to 15000 psi (1,034 bar)
Inlet/outlet connection: Female/Female 10 to 32
Port geometry: Parker (1/16 CPI)
Wrench flat: ³/₈"