Freezer boxes (Cryoboxes), True North®
Leverandør: HEATHROW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HS120371 HS120372 HS120373 HS120376 HS120388 HS120392 HS120367
HEATHS120371EA 691 DKK
HEATHS120371 HEATHS120372 HEATHS120373 HEATHS120376 HEATHS120388 HEATHS120392 HEATHS120367
Freezer boxes (Cryoboxes), True North®
Bokse Æsker til Kryorør
True North® freezer boxes (cryoboxes) were designed with sustainability in mind and are made from a specially formulated corrugated or thin film polypropylene.
  • Easy self-assemble boxes shipped flat-packed
  • More durable than card boxes as they are not susceptible to moisture
  • Dividers included with the boxes
  • Mold resistant
  • Store down to temperatures of –80 °C (Corrugated PP boxes)/–196 °C (Thin PP film boxes)
Boxes are moisture resistant, which significantly reduces the chance of mold growth. With superior durability, polypropylene outlasts the cardboard box alternatives that are susceptible to softening and producing dust and can freeze (down to temperatures of –80 °C) and thaw multiple times without a problem.
Thin PP film boxes can be used in mechanical freezers and vessels down to 196 °C (Vapour Phase only).